• USA Home
  • C7125 - β-Carbonaphthoxycholine iodide

EMAIL THIS PAGE TO A FRIEND
C7125 Sigma-Aldrich

β-Carbonaphthoxycholine iodide

Purchase

Properties

storage temp.   −20°C
SMILES string   I.C[N](C)(C)CCOC(=O)Oc1ccc2ccccc2c1

Description

Substrates

A cholinesterase substrate

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?