EMAIL THIS PAGE TO A FRIEND
C7128 Sigma-Aldrich

Chlorotrianisene

~95%

Synonym: TACE

Purchase

Properties

assay   ~95%
SMILES string   COc1ccc(cc1)C(\Cl)=C(/c2ccc(OC)cc2)c3ccc(OC)cc3
InChI   1S/C23H21ClO3/c1-25-19-10-4-16(5-11-19)22(17-6-12-20(26-2)13-7-17)23(24)18-8-14-21(27-3)15-9-18/h4-15H,1-3H3
InChI key   BFPSDSIWYFKGBC-UHFFFAOYSA-N
Gene Information   human ... ESR2(2100)

Description

Biochem/physiol Actions

Non-steroidal estrogenic compound which is metabolized by liver microsomal enzymes primarily to the mono-O-demethylated derivative.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
RTECS 
KV0600000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?