EMAIL THIS PAGE TO A FRIEND
C7130 Sigma-Aldrich

Z-Lys-OH

Synonym: Nα-(Carbobenzyloxy)-L-lysine, Nα-Cbz-L-lysine, Nα-Z-L-Lysine

  • CAS Number 2212-75-1

  • Linear Formula NH2(CH2)4CH(NHCOOCH2C6H5)COOH

  • Molecular Weight 280.32

  •  Beilstein/REAXYS Number 2153826

  •  EC Number 218-662-6

  •  MDL number MFCD00038204

Purchase

Properties

mp   226-231 °C (dec.) (lit.)
storage temp.   −20°C
SMILES string   NCCCC[C@H](NC(=O)OCc1ccccc1)C(O)=O
InChI   1S/C14H20N2O4/c15-9-5-4-8-12(13(17)18)16-14(19)20-10-11-6-2-1-3-7-11/h1-3,6-7,12H,4-5,8-10,15H2,(H,16,19)(H,17,18)/t12-/m0/s1
InChI key   OJTJKAUNOLVMDX-LBPRGKRZSA-N

Description

Application

Educt for the convenient preparation of L-α-aminoadipic acid.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

359793 Z-Lys-OH, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?