• USA Home
  • C7131 - Z-Leu-Leu methyl ester

EMAIL THIS PAGE TO A FRIEND
C7131 Sigma-Aldrich

Z-Leu-Leu methyl ester

  • CAS Number 3504-37-8

  • Empirical Formula (Hill Notation) C21H32N2O5

  • Molecular Weight 392.49

  •  MDL number MFCD00026492

  •  PubChem Substance ID 24892936

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
form   solid
application(s)   cell analysis: suitable (proteosome inhibitor)
storage temp.   −20°C
SMILES string   CC(C)C[C@@H](C(N[C@H](C(OC)=O)CC(C)C)=O)NC(OCC1=CC=CC=C1)=O
InChI   1S/C21H32N2O5/c1-14(2)11-17(19(24)22-18(12-15(3)4)20(25)27-5)23-21(26)28-13-16-9-7-6-8-10-16/h6-10,14-15,17-18H,11-13H2,1-5H3,(H,22,24)(H,23,26)/t17-,18-/m0/s1
InChI key   FYKHRTVYUNLJLJ-ROUUACIJSA-N

Description

Amino Acid Sequence

Z-Leu-Leu-Omet

Biochem/physiol Actions

N-CBZ-Ieu-Ieu methyl ester, an N-terminal and C-terminal blocked dipeptide, and related molecules may be useful in analysis of proteosome proteases.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?