EMAIL THIS PAGE TO A FRIEND
C7139 Sigma-Aldrich

Chromosorb® G

Synonym: Diatomaceous earth, flux-calcined

Purchase

Properties

InChI Key   VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Description

Legal Information

Chromosorb is a registered trademark of Imerys Minerals California, Inc.

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?