EMAIL THIS PAGE TO A FRIEND
C7238 Sigma-Aldrich

CV-3988 hydrate

≥98% (HPLC)

Synonym: (±)-[3-(N-Octadecylcarbamoyloxy)-2-methoxy]propyl 2-thiazolioethyl phosphate hydrate

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, CI-CZ, Cell Biology, Cell Signaling and Neuroscience,
InChI Key   QVRNNMBNYYSVGY-UHFFFAOYSA-N
assay   ≥98% (HPLC)
form   powder
storage condition   desiccated
color   off-white
solubility   ethanol: 10 mg/mL, clear (warmed)
storage temp.   −20°C
Gene Information   human ... PTAFR(5724)

Description

Packaging

5 mg in glass bottle

Biochem/physiol Actions

Competitive PAF receptor antagonist. Inhibits PAF-induced human platelet aggregation (3-30 μM) and bronchoconstriction in the guinea pig.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?