EMAIL THIS PAGE TO A FRIEND
C7276 Sigma

Z-Dehydro-Ala-OMe

Synonym: N-Cbz-Dehydroalanine methyl ester, Z-Dehydroalanine methyl ester

  • CAS Number 21149-17-7

  • Linear Formula C6H5CH2O2CNHC(=CH2)CO2CH3

  • Molecular Weight 235.24

  •  Beilstein Registry Number 2056075

  •  MDL number MFCD00056245

Purchase

Properties

InChI Key   STFUIEDYPRMRNN-UHFFFAOYSA-N
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

96077 Z-Dehydro-Ala-OMe, ≥98.0% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?