• USA Home
  • C7393 - L-Carnitine inner salt

EMAIL THIS PAGE TO A FRIEND
C7393 Sigma-Aldrich

L-Carnitine inner salt

from equine muscle, ~98%

Synonym: (−)-(R)-3-Hydroxy-4-(trimethylammonio)butyrate, Vitamin BT

  • CAS Number 541-15-1

  • Linear Formula (CH3)3N+CH2CH(OH)CH2COO-

  • Molecular Weight 161.20

  •  Beilstein/REAXYS Number 4292315

  •  EC Number 208-768-0

  •  MDL number MFCD00038747

Purchase

Properties

biological source   equine muscle
assay   ~98%
mp   197-212 °C (lit.)
storage temp.   −20°C
SMILES string   C[N+](C)(C)C[C@H](O)CC([O-])=O
InChI   1S/C7H15NO3/c1-8(2,3)5-6(9)4-7(10)11/h6,9H,4-5H2,1-3H3/t6-/m1/s1
InChI key   PHIQHXFUZVPYII-ZCFIWIBFSA-N

Description

Biochem/physiol Actions

Carnitine is a quaternary amine that occurs naturally in most mammalian tissue. It is present in relatively high concentrations in skeletal muscle and heart where it is involved in regulating energy metabolism. It shifts glucose metabolism from glycolysis to glycogen storage and enhances the transport of long chain fatty acids into the mitochondria where they are oxidized for energy production.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
RTECS 
BP2980000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

C0158 L-Carnitine inner salt, synthetic, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?