• USA Home
  • C7652 - Calcium nitrate tetrahydrate

EMAIL THIS PAGE TO A FRIEND
C7652 Sigma-Aldrich

Calcium nitrate tetrahydrate

suitable for cell culture

Purchase

Properties

InChI Key   ICSSIKVYVJQJND-UHFFFAOYSA-N
application(s)   cell culture | mammalian: suitable
mp   44 °C (dec.) (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
EW3000000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?