• USA Home
  • C7705 - Calcium bromide hydrate

EMAIL THIS PAGE TO A FRIEND
C7705 Sigma-Aldrich

Calcium bromide hydrate

≥98%

Purchase

Properties

InChI Key   NQMKEZBQFUTOQC-UHFFFAOYSA-L
assay   ≥98%
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?