EMAIL THIS PAGE TO A FRIEND
C7876 Sigma-Aldrich

Z-Gly-Ala

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
storage temp.   −20°C
SMILES string   O=C(N[C@@H](C)C(O)=O)CNC(OCC1=CC=CC=C1)=O
InChI   1S/C13H16N2O5/c1-9(12(17)18)15-11(16)7-14-13(19)20-8-10-5-3-2-4-6-10/h2-6,9H,7-8H2,1H3,(H,14,19)(H,15,16)(H,17,18)/t9-/m0/s1
InChI key   CCIBGDNXMPNUHL-VIFPVBQESA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?