EMAIL THIS PAGE TO A FRIEND
C8001 Sigma-Aldrich

Z-Gly-Gly

  • CAS Number 2566-19-0

  • Empirical Formula (Hill Notation) C12H14N2O5

  • Molecular Weight 266.25

  •  MDL number MFCD00037783

  •  PubChem Substance ID 24893004

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?