• USA Home
  • C8386 - β-Chloro-L-alanine hydrochloride

EMAIL THIS PAGE TO A FRIEND
C8386

β-Chloro-L-alanine hydrochloride

95-97%

  • CAS Number 51887-89-9

  • Empirical Formula (Hill Notation) C3H6ClNO2 · HCl

  • Molecular Weight 160.00

  •  MDL number MFCD00070398

Purchase

Properties

assay   95-97%
storage temp.   −20°C
SMILES string   Cl[H].N[C@@H](CCl)C(O)=O
InChI   1S/C3H6ClNO2.ClH/c4-1-2(5)3(6)7;/h2H,1,5H2,(H,6,7);1H/t2-;/m0./s1
InChI key   IENJPSDBNBGIEL-DKWTVANSSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?