• USA Home
  • C8794 - 1α,2α[N]-3H-Cholesterol

EMAIL THIS PAGE TO A FRIEND
C8794 Sigma-Aldrich

1α,2α[N]-3H-Cholesterol

ethanol solution

Purchase

Properties

form   ethanol solution
extent of labeling   40-60 Ci/mmol
storage temp.   2-8°C
SMILES string   [H]C1C([H])[C@]2(C)C(C[C@@H]1O)=CC[C@]3([H])[C@@]4([H])CC[C@@]([H])([C@@H](C)CCCC(C)C)[C@@]4(C)CC[C@@]23[H]

Description

Packaging

serum bottle

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
1
Flash Point(F) 
57 °F
Flash Point(C) 
14 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?