• USA Home
  • C9250 - 2-Carboxyphenyl phosphate

EMAIL THIS PAGE TO A FRIEND
C9250 Supelco

2-Carboxyphenyl phosphate

Synonym: 2-Phosphonooxybenzoic acid, Fosfosal, Salicylic acid phosphate

  • CAS Number 6064-83-1

  • Empirical Formula (Hill Notation) C7H7O6P

  • Molecular Weight 218.10

  •  Beilstein/REAXYS Number 2619142

  •  EC Number 227-993-5

  •  MDL number MFCD00042645

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
VO2770000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?