EMAIL THIS PAGE TO A FRIEND
C9376 Sigma-Aldrich

Z-Gly-Gly-Val

  • CAS Number 34152-86-8

  • Empirical Formula (Hill Notation) C17H23N3O6

  • Molecular Weight 365.38

  •  MDL number MFCD00056144

  •  PubChem Substance ID 24893152

Purchase

Properties

storage temp.   −20°C
SMILES string   CC(C)C(NC(=O)CNC(=O)CNC(=O)OCc1ccccc1)C(O)=O

Description

Amino Acid Sequence

Z-GLY-GLY-VAL

Application

Cbz-Gly-Gly-Val (Cbz-GGV), an N-terminal carboxybenzyl blocked tripeptide, may be use in solid phase peptide synthesis (SPPS).

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?