• USA Home
  • C9403 - 5′-Chloro-5′-deoxycytidine

EMAIL THIS PAGE TO A FRIEND
C9403 Sigma-Aldrich

5′-Chloro-5′-deoxycytidine

  • CAS Number 31652-78-5

  • Empirical Formula (Hill Notation) C9H12ClN3O4

  • Molecular Weight 261.66

Purchase

Properties

storage temp.   2-8°C
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?