EMAIL THIS PAGE TO A FRIEND
C9548 Sigma-Aldrich

Coniferyl alcohol

~98% (GC)

Synonym: 3-(4-Hydroxy-3-methoxyphenyl)-2-propen-1-ol, 4-Hydroxy-3-methoxycinnamyl alcohol

  • CAS Number 458-35-5

  • Linear Formula HOC6H3(OCH3)CH=CHCH2OH

  • Molecular Weight 180.20

  •  Beilstein/REAXYS Number 2048961

  •  EC Number 207-277-9

  •  MDL number MFCD00002922

Purchase

Properties

InChI Key   JMFRWRFFLBVWSI-NSCUHMNNSA-N
assay   ~98% (GC)
bp   163-165 °C/3 mmHg (lit.)
mp   75-80 °C (lit.)
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

223735 Coniferyl alcohol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?