• USA Home
  • C9655 - (−)-Camphor-10-sulfonic acid

EMAIL THIS PAGE TO A FRIEND
C9655 Sigma-Aldrich

(−)-Camphor-10-sulfonic acid

Synonym: (1R)-(−)-Camphor-10-sulfonic acid

  • CAS Number 35963-20-3

  • Empirical Formula (Hill Notation) C10H16O4S

  • Molecular Weight 232.30

  •  Beilstein/REAXYS Number 2809676

  •  EC Number 252-817-9

  •  MDL number MFCD00064158

Purchase

Properties

storage temp.   2-8°C
SMILES string   CC1(C)C2CC[C@]1(CS(O)(=O)=O)C(=O)C2
InChI   1S/C10H16O4S/c1-9(2)7-3-4-10(9,8(11)5-7)6-15(12,13)14/h7H,3-6H2,1-2H3,(H,12,13,14)/t7-,10-/m0/s1
InChI key   MIOPJNTWMNEORI-XVKPBYJWSA-N
Safety & Documentation

Safety Information

RIDADR 
UN 3261 8/PG 2
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

282146 (1R)-(−)-10-Camphorsulfonic acid, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?