• USA Home
  • D031 - N,N-Dipropyldopamine hydrobromide

EMAIL THIS PAGE TO A FRIEND
D031 Supelco

N,N-Dipropyldopamine hydrobromide

solid

  • CAS Number 65273-66-7

  • Empirical Formula (Hill Notation) C14H23NO2 · HBr

  • Molecular Weight 318.25

  •  MDL number MFCD00069242

Purchase

Properties

form   solid
color   white
solubility   aqueous base: unstable
  ethanol: soluble (Solutions should be freshly prepared and kept at 4 °C.)
  oxygen-free water containing 0.1% sodium metabisulfite or other antioxidant: soluble (Solutions should be freshly prepared and kept at 4 °C.)
storage temp.   2-8°C
Gene Information   human ... DRD1(1812), DRD2(1813), DRD3(1814), DRD4(1815), DRD5(1816)

Description

Biochem/physiol Actions

Dopamine receptor agonist.

Packaging

Packaged under argon.

Caution

Photosensitive

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
RB5962000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?