• USA Home
  • D0630 - DNP-DL-methionine sulfone

EMAIL THIS PAGE TO A FRIEND
D0630 Sigma-Aldrich

DNP-DL-methionine sulfone

Synonym: N-(2,4-Dinitrophenyl)-DL-methionine sulfone

  • CAS Number 16068-18-1

  • Empirical Formula (Hill Notation) C11H13N3O8S

  • Molecular Weight 347.30

  •  MDL number MFCD00057623

  •  PubChem Substance ID 24893249

Purchase

Properties

Related Categories Amino Acid Analysis, Amino Acid Analysis Reagents, Biochemicals and Reagents, Peptide Analysis and Characterization, Peptides More...
storage temp.   −20°C
SMILES string   CS(=O)(=O)CCC(Nc1ccc(cc1N(=O)=O)N(=O)=O)C(O)=O
InChI   1S/C11H13N3O8S/c1-23(21,22)5-4-9(11(15)16)12-8-3-2-7(13(17)18)6-10(8)14(19)20/h2-3,6,9,12H,4-5H2,1H3,(H,15,16)
InChI key   KFPFJKADCNQPPQ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?