• USA Home
  • D0650 - 6,15-Diketo-13,14-dihydro-prostaglandin F

EMAIL THIS PAGE TO A FRIEND
D0650 Sigma-Aldrich

6,15-Diketo-13,14-dihydro-prostaglandin F

Purchase

Properties

shipped in   dry ice
storage temp.   −20°C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?