EMAIL THIS PAGE TO A FRIEND
D0775 Sigma

1,6-Dinitropyrene

  • CAS Number 42397-64-8

  • Empirical Formula (Hill Notation) C16H8N2O4

  • Molecular Weight 292.25

  •  MDL number MFCD00010494

Purchase

Properties

InChI Key   GUXACCKTQWVTLG-UHFFFAOYSA-N
mp   >300 °C (lit.)
storage temp.   −20°C
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UR2455000
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?