• USA Home
  • D0881 - N-Dansyl-DL-threonine cyclohexylammonium salt

EMAIL THIS PAGE TO A FRIEND
D0881 Sigma-Aldrich

N-Dansyl-DL-threonine cyclohexylammonium salt

  • CAS Number 84540-66-9

  • Empirical Formula (Hill Notation) C16H20N2O5S · C6H13N

  • Molecular Weight 451.58

  •  EC Number 283-161-1

  •  PubChem Substance ID 329798647

Purchase
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?