EMAIL THIS PAGE TO A FRIEND
D1005 Sigma-Aldrich

DNP-DL-norvaline

Synonym: N-(2,4-Dinitrophenyl)-DL-norvaline

  • CAS Number 31356-37-3

  • Empirical Formula (Hill Notation) C11H13N3O6

  • Molecular Weight 283.24

  •  MDL number MFCD00049235

  •  PubChem Substance ID 24893313

Purchase

Properties

Related Categories Amino Acid Analysis, Amino Acid Analysis Reagents, Biochemicals and Reagents, Peptide Analysis and Characterization, Peptides More...
storage temp.   −20°C
SMILES string   CCCC(Nc1ccc(cc1N(=O)=O)N(=O)=O)C(O)=O
InChI   1S/C11H13N3O6/c1-2-3-9(11(15)16)12-8-5-4-7(13(17)18)6-10(8)14(19)20/h4-6,9,12H,2-3H2,1H3,(H,15,16)
InChI key   WSJNAXWRYWZIBW-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?