EMAIL THIS PAGE TO A FRIEND
D1035 Sigma-Aldrich

7-Deazaguanine

~95%, crystalline

Synonym: 2-Amino-4-hydroxy­pyrrolo­[2,3-d]­pyrimidine

  • CAS Number 7355-55-7

  • Empirical Formula (Hill Notation) C6H6N4O

  • Molecular Weight 150.14

  •  MDL number MFCD00079103

  •  PubChem Substance ID 24893330

Purchase

Properties

assay   ~95%
form   crystalline
color   tan
SMILES string   Nc1nc(O)c2cc[nH]c2n1

Description

Application

Ethidium bromide efficiently intercalates into synthetic duplex DNAs with 7-deazaguanine in place of guanine, but shows none of the fluorescence enhancement usually observed upon intercalation. 7-Deazaadenine-duplexes show about 70% of the expected fluorescence enhancement.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Indoles, Purines, and Their Isosteres

Substituted indoles and purines have frequently been referred to as “privileged structures” since they are capable of binding to multiple receptors with high affinity, and thus have applications acro...
Keywords: Building blocks, Medicinal chemistry

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?