• USA Home
  • D1056 - N2,N2-Dimethylguanosine

EMAIL THIS PAGE TO A FRIEND
D1056 Sigma-Aldrich

N2,N2-Dimethylguanosine

  • CAS Number 2140-67-2

  • Empirical Formula (Hill Notation) C12H17N5O5

  • Molecular Weight 311.29

  •  MDL number MFCD00057049

Purchase

Properties

storage temp.   2-8°C
SMILES string   CN(C)C1=Nc2c(ncn2[C@@H]3N[C@H](CO)[C@@H](O)[C@H]3O)C(=O)N1

Description

Application

Proposed as an indicator for tRNA half-life in normal vs. cancerous tissues.

Other Notes

Modified nucleoside found in tRNA and snRNA of archaebacteria and eukaryotes.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?