• USA Home
  • D1130 - Nδ-DNP-L-ornithine hydrochloride

EMAIL THIS PAGE TO A FRIEND
D1130 Sigma-Aldrich

Nδ-DNP-L-ornithine hydrochloride

Synonym: Nδ-(2,4-Dinitrophenyl)-L-ornithine hydrochloride

  • CAS Number 64821-45-0

  • Empirical Formula (Hill Notation) C11H14N4O6 · HCl

  • Molecular Weight 334.71

  •  MDL number MFCD00039053

  •  PubChem Substance ID 24893358

Purchase

Properties

Related Categories Amino Acid Analysis, Amino Acid Analysis Reagents, Biochemicals and Reagents, Peptide Analysis and Characterization, Peptides More...
storage temp.   −20°C
SMILES string   Cl.NC(CCCNc1ccc(cc1N(=O)=O)N(=O)=O)C(O)=O
InChI   1S/C11H14N4O6.ClH/c12-8(11(16)17)2-1-5-13-9-4-3-7(14(18)19)6-10(9)15(20)21;/h3-4,6,8,13H,1-2,5,12H2,(H,16,17);1H
InChI key   VJBDFLCZEZSODJ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?