EMAIL THIS PAGE TO A FRIEND
D1138 Sigma-Aldrich

3′-Deoxythymidine

Synonym: 2′,3′-Dideoxythymidine, DDT

  • CAS Number 3416-05-5

  • Empirical Formula (Hill Notation) C10H14N2O4

  • Molecular Weight 226.23

  •  Beilstein/REAXYS Number 21884

  •  MDL number MFCD00010570

Purchase

Properties

mp   155-156 °C (lit.)
storage temp.   −20°C
SMILES string   CC1=CN([C@H]2CC[C@@H](CO)O2)C(=O)NC1=O
InChI   1S/C10H14N2O4/c1-6-4-12(10(15)11-9(6)14)8-3-2-7(5-13)16-8/h4,7-8,13H,2-3,5H2,1H3,(H,11,14,15)/t7-,8+/m0/s1
InChI key   XKKCQTLDIPIRQD-JGVFFNPUSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

323616 3′-Deoxythymidine, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?