• USA Home
  • D132 - N,N-Dipropyl-5-carboxamidotryptamine maleate salt

EMAIL THIS PAGE TO A FRIEND
D132 Sigma-Aldrich

N,N-Dipropyl-5-carboxamidotryptamine maleate salt

solid

  • CAS Number 74885-25-9

  • Empirical Formula (Hill Notation) C17H25N3O · C4H4O4

  • Molecular Weight 403.47

  •  MDL number MFCD00069264

  •  PubChem Substance ID 24278402

Purchase

Properties

form   solid
color   light yellow
solubility   0.1 M NaOH: soluble
  DMSO: soluble
  H2O: soluble
  aqueous acid: unstable
  ethanol: soluble
Gene Information   human ... HTR1A(3350)

Description

Biochem/physiol Actions

Potent and selective 5-HT1A serotonin receptor agonist.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?