EMAIL THIS PAGE TO A FRIEND
D135 Sigma-Aldrich

Domoic acid

  • CAS Number 14277-97-5

  • Empirical Formula (Hill Notation) C15H21NO6

  • Molecular Weight 311.33

  •  Beilstein/REAXYS Number 46376

  •  MDL number MFCD06795841

Purchase

Properties

storage temp.   −20°C
SMILES string   OC([C@H](C)/C=C/C=C([C@@H]1[C@H](CC(O)=O)[C@@H](C(O)=O)NC1)/C)=O
InChI   1S/C15H21NO6/c1-8(4-3-5-9(2)14(19)20)11-7-16-13(15(21)22)10(11)6-12(17)18/h3-5,9-11,13,16H,6-7H2,1-2H3,(H,17,18)(H,19,20)(H,21,22)/b5-3+,8-4-/t9-,10+,11-,13+/m1/s1
InChI key   VZFRNCSOCOPNDB-AOKDLOFSSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UX9665100

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

D6152 Domoic acid, ≥90% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?