EMAIL THIS PAGE TO A FRIEND
D146 Sigma-Aldrich

Debrisoquine sulfate

Synonym: 3,4-Dihydro-2(1H)-isoquinolinecarboximidamide, Debrisoquin sulfate, Ro 5-33071

Purchase

Properties

InChI Key   CAYGYVYWRIHZCQ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
NW7010000

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

D1306 Debrisoquine sulfate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?