EMAIL THIS PAGE TO A FRIEND
D157 Sigma-Aldrich

Dexamethasone

Synonym: (11β,16α)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-16α-methylprednisolone, Prednisolone F

  • CAS Number 50-02-2

  • Empirical Formula (Hill Notation) C22H29FO5

  • Molecular Weight 392.46

  •  Beilstein/REAXYS Number 2066651

  •  EC Number 200-003-9

  •  MDL number MFCD00064136

Purchase

Properties

InChI Key   UREBDLICKHMUKA-CXSFZGCWSA-N
mp   262-264 °C (lit.)

Description

Biochem/physiol Actions

Glucocorticoid anti-inflammatory agent. Regulates T cell survival, growth, and differentiation. Inhibits the induction of nitric oxide synthase.

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
TU3980000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D1756 Dexamethasone, ≥98% (HPLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?