• USA Home
  • D1784 - Dodecylethyldimethylammonium bromide

EMAIL THIS PAGE TO A FRIEND
D1784 Sigma-Aldrich

Dodecylethyldimethylammonium bromide

Synonym: Dimethyldodecylethylammonium bromide, Dodecyldimethylethylammonium bromide

  • CAS Number 68207-00-1

  • Linear Formula [CH3(CH2)11N+(CH3)2C2H5]Br-

  • Molecular Weight 322.37

  •  Beilstein/REAXYS Number 6007021

  •  EC Number 269-249-2

  •  MDL number MFCD00043170

  •  PubChem Substance ID 24893542

Purchase

Properties

description   cationic
mp   185-188 °C (lit.)
SMILES string   [Br-].CCCCCCCCCCCC[N+](C)(C)CC
InChI   1S/C16H36N.BrH/c1-5-7-8-9-10-11-12-13-14-15-16-17(3,4)6-2;/h5-16H2,1-4H3;1H/q+1;/p-1
InChI key   FFGSPQDSOUPWGY-UHFFFAOYSA-M
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

44165 Dodecylethyldimethylammonium bromide, ≥98.0% (AT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?