• USA Home
  • D1897 - 5,6-Dihydrodeoxyuridine

EMAIL THIS PAGE TO A FRIEND
D1897 Sigma-Aldrich

5,6-Dihydrodeoxyuridine

  • CAS Number 5626-99-3

  • Empirical Formula (Hill Notation) C9H14N2O5

  • Molecular Weight 230.22

  •  MDL number MFCD00070301

  •  PubChem Substance ID 24893567

Purchase

Properties

form   solid
storage temp.   −20°C
SMILES string   OCC1OC(CC1O)N2CCC(=O)NC2=O
InChI   1S/C9H14N2O5/c12-4-6-5(13)3-8(16-6)11-2-1-7(14)10-9(11)15/h5-6,8,12-13H,1-4H2,(H,10,14,15)
InChI key   XMJRLEURHMTTRX-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?