• USA Home
  • D2134 - 2′-Deoxycytidine 3′-monophosphate

EMAIL THIS PAGE TO A FRIEND
D2134 Sigma-Aldrich

2′-Deoxycytidine 3′-monophosphate

95-98%

  • CAS Number 6220-63-9

  • Empirical Formula (Hill Notation) C9H14N3O7P

  • Molecular Weight 307.20

  •  MDL number MFCD00070165

Purchase

Properties

assay   95-98%
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?