• USA Home
  • D2279 - Nα,Nε-Diacetyl-Lys-D-Ala-D-lactic acid

EMAIL THIS PAGE TO A FRIEND
D2279 Sigma-Aldrich

Nα,Nε-Diacetyl-Lys-D-Ala-D-lactic acid

  • CAS Number 65882-12-4

  • Empirical Formula (Hill Notation) C16H27N3O7

  • Molecular Weight 373.40

  •  MDL number MFCD00038705

  •  PubChem Substance ID 24893674

Purchase

Description

Substrates

Substrate for penicillin-sensitive D-alanine carboxypeptidase.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?