• USA Home
  • D2657 - P1,P4-Di(adenosine-5′) tetraphosphate, periodate oxidized sodium salt

EMAIL THIS PAGE TO A FRIEND
D2657 Sigma-Aldrich

P1,P4-Di(adenosine-5′) tetraphosphate, periodate oxidized sodium salt

90-95%

Synonym: P1,P4-Di(adenosine-2′,3′-dialdehyde 5′-tetraphosphate

  • CAS Number 112968-03-3

  • Empirical Formula (Hill Notation) C20H24N10O19P4

  • Molecular Weight 832.36

  •  MDL number MFCD00057982

  •  PubChem Substance ID 24893704

Purchase

Properties

assay   90-95%
storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?