• USA Home
  • D2772 - Dimethylamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
D2772 Sigma-Aldrich

Dimethylamine hydrochloride

>99% (titration)

  • CAS Number 506-59-2

  • Linear Formula (CH3)2NH · HCl

  • Molecular Weight 81.54

  •  Beilstein/REAXYS Number 3589311

  •  EC Number 208-046-5

  •  MDL number MFCD00012477

Purchase

Properties

assay   >99% (titration)
mp   170-173 °C (lit.)
SMILES string   Cl[H].CNC
InChI   1S/C2H7N.ClH/c1-3-2;/h3H,1-2H3;1H
InChI key   IQDGSYLLQPDQDV-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
IQ0220000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

126365 Dimethylamine hydrochloride, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?