• USA Home
  • D3258 - 2,3-Diphospho-D-glyceric acid tris salt

EMAIL THIS PAGE TO A FRIEND
D3258 Sigma

2,3-Diphospho-D-glyceric acid tris salt

~98%

Synonym: D-Glycerate 2,3-diphosphate tris salt

  • CAS Number 102814-02-8

  • Empirical Formula (Hill Notation) C3H8O10P2

  • Molecular Weight 266.04

  •  MDL number MFCD00070076

Purchase

Properties

assay   ~98%
impurities   3-phosphoglyceric acid, essentially free
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

D9134 2,3-Diphospho-D-glyceric acid penta(cyclohexylammonium) salt, ≥97% (TLC)
D5764 2,3-Diphospho-D-glyceric acid pentasodium salt, glycolysis metabolite

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?