EMAIL THIS PAGE TO A FRIEND
D3275 Sigma-Aldrich

S-Decylglutathione

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides and Tripeptides, Glutathione and Derivatives, Peptides
InChI Key   XAAOGZMKEJNHLW-UHFFFAOYSA-N
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?