• USA Home
  • D3526 - 1-Deoxy-1-nitro-D-sorbitol

EMAIL THIS PAGE TO A FRIEND
D3526 Sigma-Aldrich

1-Deoxy-1-nitro-D-sorbitol

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates D-F, Monosaccharide More...
InChI Key   HOFCJTOUEGMYBT-SLPGGIOYSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?