• USA Home
  • D3532 - 1,2-Dipalmitoyl-3-myristoyl-rac-glycerol

EMAIL THIS PAGE TO A FRIEND
D3532 Sigma-Aldrich

1,2-Dipalmitoyl-3-myristoyl-rac-glycerol

~99%

Synonym: 1,2-Dihexadecanoyl-3-tetradecanoyl-rac-glycerol

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Glycerolipids, Glycerophospholipid Metabolism, Lipids,
InChI Key   JWVXCFSNEOMSHH-UHFFFAOYSA-N
assay   ~99%
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Milli-Q® Water Purification Solutions
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?