• USA Home
  • D3885 - 2,4-Diamino-6,7-diisopropylpteridine phosphate salt

EMAIL THIS PAGE TO A FRIEND
D3885

2,4-Diamino-6,7-diisopropylpteridine phosphate salt

Purchase

Properties

mp   232-234 °C (lit.)
storage temp.   2-8°C
SMILES string   OP(O)(O)=O.CC(C)c1nc2nc(N)nc(N)c2nc1C(C)C
InChI   1S/C12H18N6.H3O4P/c1-5(2)7-8(6(3)4)16-11-9(15-7)10(13)17-12(14)18-11;1-5(2,3)4/h5-6H,1-4H3,(H4,13,14,16,17,18);(H3,1,2,3,4)
InChI key   YNQBQYASRYRNRY-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

D0781 2,4-Diamino-6,7-diisopropylpteridine phosphate salt, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?