• USA Home
  • D4030 - N6,3′-O-Dibenzoyl-2′-deoxyadenosine

EMAIL THIS PAGE TO A FRIEND
D4030 Sigma-Aldrich

N6,3′-O-Dibenzoyl-2′-deoxyadenosine

≥98%

Purchase

Properties

assay   ≥98%
storage temp.   −20°C
SMILES string   OCC1OC(CC1OC(=O)c2ccccc2)n3cnc4c(NC(=O)c5ccccc5)ncnc34
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?