• USA Home
  • D4067 - Delta Sleep Inducing Peptide

EMAIL THIS PAGE TO A FRIEND
D4067 Sigma-Aldrich

Delta Sleep Inducing Peptide

≥97% (HPLC)

Synonym: DSIP, Emideltide, Sleep inducing peptide, Trp-Ala-Gly-Gly-Asp-Ala-Ser-Gly-Glu

  • CAS Number 62568-57-4

  • Empirical Formula (Hill Notation) C35H48N10O15

  • Molecular Weight 848.81

  •  MDL number MFCD00076883

Purchase

Properties

InChI Key   ZRZROXNBKJAOKB-GFVHOAGBSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Amino Acid Sequence

Trp-Ala-Gly-Gly-Asp-Ala-Ser-Gly-Glu-OH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?