EMAIL THIS PAGE TO A FRIEND
D4383 Sigma-Aldrich

6-Dehydroestrone

≥98%

Synonym: 1,3,5(10),6-Estra­tetra­en-3-ol-17-one, 3-Hydroxy-1,3,5(10),6-estra­tetra­en-17-one

  • CAS Number 2208-12-0

  • Empirical Formula (Hill Notation) C18H20O2

  • Molecular Weight 268.35

  •  MDL number MFCD00056393

  •  PubChem Substance ID 24893865

Purchase

Properties

assay   ≥98%
form   powder
color   white to off-white
SMILES string   [H][C@]12CC[C@]3(C)C(=O)CC[C@@]3([H])[C@]1([H])C=Cc4cc(O)ccc24
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?