• USA Home
  • D4899 - Dynorphin A Porcine Fragment 1-8

EMAIL THIS PAGE TO A FRIEND
D4899 Sigma-Aldrich

Dynorphin A Porcine Fragment 1-8

≥97% (HPLC)

  • CAS Number 75790-53-3

  • Empirical Formula (Hill Notation) C46H72N14O10

  • Molecular Weight 981.15

  •  MDL number MFCD00076357

  •  PubChem Substance ID 24893905

Purchase

Properties

assay   ≥97% (HPLC)
composition   Peptide content, ~70%
storage temp.   −20°C
Gene Information   human ... OPRD1(4985), OPRK1(4986), OPRM1(4988)
pig ... PDYN(445529)
rat ... Oprd1(24613), Oprm1(25601)

Description

Amino Acid Sequence

Tyr-Gly-Gly-Phe-Leu-Arg-Arg-Ile

Biochem/physiol Actions

Potent κ opioid agonist; its agonist activity at μ and δ receptors may be due to hydrolysis of the peptide by proteases in the membrane or cell preparations.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?