EMAIL THIS PAGE TO A FRIEND
D5875 Sigma

2′-Deoxyadenosine

98-100%

Synonym: 9-(2-Deoxy-β-D-ribofuranosyl)adenine, Adenine deoxyriboside

  • CAS Number 16373-93-6

  • Empirical Formula (Hill Notation) C10H13N5O3 · H2O

  • Molecular Weight 269.26

  •  Beilstein/REAXYS Number 91015

  •  EC Number 213-488-7

  •  MDL number MFCD00149364

Purchase

Properties

assay   98-100%
mp   185-187 °C (dec.) (lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
AU7358600

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

D7400 2′-Deoxyadenosine monohydrate, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?