• USA Home
  • D6017 - DNP-L-cysteic acid disodium salt hydrate

EMAIL THIS PAGE TO A FRIEND
D6017 Sigma-Aldrich

DNP-L-cysteic acid disodium salt hydrate

90-95%

Synonym: N-(2,4-Dinitrophenyl)-L-cysteic acid disodium salt hydrate

  • CAS Number 16068-14-7

  • Empirical Formula (Hill Notation) C9H9N3O9S · 2Na · H2O

  • Molecular Weight 399.24

  •  PubChem Substance ID 24893993

Purchase

Properties

assay   90-95%
storage temp.   −20°C
SMILES string   [Na+].[Na+].[H]O[H].[O-]C(=O)[C@H](CS([O-])(=O)=O)Nc1ccc(cc1N(=O)=O)N(=O)=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?